The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290331.1 | NP_001277260.1 | 16 - 17 | 1362 | 152 |
NM_024898.3 | NP_079174.2 | 18 - 19 | 1471 | 152 | |
XM_006722905.2 | XP_006722968.2 | 16 - 17 | 1215 | 152 | |
XM_006722906.3 | XP_006722969.1 | 18 - 19 | 1400 | 152 | |
XM_011528318.2 | XP_011526620.1 | 17 - 18 | 1391 | 152 | |
GenBank mRNA | AK026410.1 | - | 18 - 19 | 1450 | 152 |
AK055820.1 | - | 7 - 8 | 1078 | 152 | |
AK225623.1 | - | 18 - 19 | 1450 | 152 | |
AK303226.1 | - | 16 - 17 | 1341 | 152 | |
AL713770.1 | - | 16 - 17 | 1591 | 152 | |
AL834293.1 | - | 1 - 2 | 72 | 152 | |
BC033437.1 | - | 18 - 19 | 1438 | 152 | |
BX647304.1 | - | 6 - 7 | 1101 | 152 |