The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290331.1 | NP_001277260.1 | 3 - 4 | 304 | 64 |
NM_024898.3 | NP_079174.2 | 5 - 6 | 413 | 64 | |
XM_006722905.2 | XP_006722968.2 | 4 - 5 | 283 | 64 | |
XM_006722906.3 | XP_006722969.1 | 5 - 6 | 342 | 64 | |
XM_011528318.2 | XP_011526620.1 | 4 - 5 | 333 | 64 | |
GenBank mRNA | AK026410.1 | - | 5 - 6 | 392 | 64 |
AK225623.1 | - | 5 - 6 | 392 | 64 | |
AK303226.1 | - | 3 - 4 | 283 | 64 | |
AK303509.1 | - | 4 - 5 | 318 | 64 | |
AL713770.1 | - | 4 - 5 | 521 | 64 | |
BC033437.1 | - | 5 - 6 | 380 | 64 | |
DC378383.1 | - | 5 - 6 | 413 | 64 |