Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HADHB
Entrez Gene ID
3032
Gene Name
hydroxyacyl-CoA dehydrogenase/3-ketoacyl-CoA thiolase/enoyl-CoA hydratase (trifunctional protein), beta subunit
Gene Aliases
ECHB, MSTP029, MTPB, TP-BETA
Chromosome Location
Chr.2: 26244748 - 26290465 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000183.2 NP_000174.1 14 - 15 1476 101
NM_001281512.1 NP_001268441.1 13 - 14 1431 101
NM_001281513.1 NP_001268442.1 15 - 16 1559 101
XM_011532803.1 XP_011531105.1 14 - 15 1394 101
XM_017003939.1 XP_016859428.1 14 - 15 1471 101
GenBank mRNA AF113209.1 - 14 - 15 1285 101
AK095278.1 - 12 - 13 2172 101
AK130072.1 - 14 - 15 1299 101
AK293142.1 - 13 - 14 1233 101
AK302322.1 - 14 - 15 1289 101
AK304455.1 - 15 - 16 1345 101
AK314455.1 - 14 - 15 1285 101
BC014572.1 - 14 - 15 1288 101
BC017564.2 - 14 - 15 1311 101
BC030824.1 - 14 - 15 1260 101
BC066963.1 - 14 - 15 1312 101
D16481.1 - 14 - 15 1271 101

View Full Product Details