The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085487.2 | NP_001078956.1 | 16 - 17 | 2045 | 153 |
XM_006710314.3 | XP_006710377.1 | 16 - 17 | 2061 | 153 | |
XM_011540573.2 | XP_011538875.1 | 17 - 18 | 2025 | 153 | |
XM_011540574.2 | XP_011538876.1 | 16 - 17 | 2293 | 153 | |
XM_017000195.1 | XP_016855684.1 | 17 - 18 | 2097 | 153 | |
XM_017000196.1 | XP_016855685.1 | 14 - 15 | 1947 | 153 | |
XM_017000197.1 | XP_016855686.1 | 12 - 13 | 1725 | 153 | |
GenBank mRNA | AB067502.1 | - | 12 - 13 | 2071 | 153 |
AK126835.1 | - | 10 - 11 | 1594 | 153 | |
AK292919.1 | - | 3 - 4 | 732 | 153 | |
BX537912.1 | - | 3 - 4 | 309 | 153 | |
BX648168.1 | - | 13 - 14 | 1867 | 153 |