Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYSM1
Entrez Gene ID
114803
Gene Name
Myb like, SWIRM and MPN domains 1
Gene Aliases
2A-DUB, 2ADUB
Chromosome Location
Chr.1: 58654739 - 58700091 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085487.2 NP_001078956.1 1 - 2 85 146
XM_006710314.3 XP_006710377.1 1 - 2 101 146
XM_011540574.2 XP_011538876.1 1 - 2 82 146
XM_017000196.1 XP_016855685.1 1 - 2 90 146
GenBank mRNA CR627323.1 - 1 - 2 83 146

View Full Product Details