The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005468.2 | NP_005459.2 | 12 - 13 | 1526 | 78 |
XM_011544707.2 | XP_011543009.1 | 13 - 14 | 1494 | 78 | |
XM_011544709.2 | XP_011543011.1 | 12 - 13 | 1866 | 78 | |
XM_011544711.2 | XP_011543013.1 | 11 - 12 | 1800 | 78 | |
XM_011544712.2 | XP_011543014.1 | 11 - 12 | 1801 | 78 | |
GenBank mRNA | AF010141.1 | - | 3 - 4 | 256 | 78 |
AJ012371.1 | - | 12 - 13 | 1526 | 78 |