Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAALADL1
Entrez Gene ID
10004
Gene Name
N-acetylated alpha-linked acidic dipeptidase-like 1
Gene Aliases
I100, NAALADASEL
Chromosome Location
Chr.11: 65044818 - 65059020 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005468.2 NP_005459.2 13 - 14 1616 80
XM_011544707.2 XP_011543009.1 14 - 15 1584 80
XM_011544711.2 XP_011543013.1 12 - 13 1890 80
GenBank mRNA AF010141.1 - 4 - 5 346 80
AJ012371.1 - 13 - 14 1616 80

View Full Product Details