The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005468.2 | NP_005459.2 | 1 - 2 | 203 | 92 |
XM_011544706.2 | XP_011543008.1 | 1 - 2 | 523 | 92 | |
XM_011544707.2 | XP_011543009.1 | 2 - 3 | 171 | 92 | |
XM_011544708.2 | XP_011543010.1 | 1 - 2 | 523 | 92 | |
XM_011544709.2 | XP_011543011.1 | 1 - 2 | 543 | 92 | |
XM_011544710.2 | XP_011543012.1 | 1 - 2 | 540 | 92 | |
XM_011544711.2 | XP_011543013.1 | 1 - 2 | 545 | 92 | |
XM_011544712.2 | XP_011543014.1 | 1 - 2 | 546 | 92 | |
GenBank mRNA | AJ012371.1 | - | 1 - 2 | 203 | 92 |