The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005468.2 | NP_005459.2 | 3 - 4 | 497 | 56 |
XM_011544706.2 | XP_011543008.1 | 3 - 4 | 817 | 56 | |
XM_011544707.2 | XP_011543009.1 | 4 - 5 | 465 | 56 | |
XM_011544708.2 | XP_011543010.1 | 3 - 4 | 817 | 56 | |
XM_011544709.2 | XP_011543011.1 | 3 - 4 | 837 | 56 | |
XM_011544710.2 | XP_011543012.1 | 3 - 4 | 834 | 56 | |
XM_011544711.2 | XP_011543013.1 | 3 - 4 | 839 | 56 | |
XM_011544712.2 | XP_011543014.1 | 3 - 4 | 840 | 56 | |
GenBank mRNA | AJ012371.1 | - | 3 - 4 | 497 | 56 |