The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143937.1 | NP_001137409.1 | 3 - 4 | 297 | 130 |
NM_002786.3 | NP_002777.1 | 3 - 4 | 297 | 130 | |
NM_148976.2 | NP_683877.1 | 4 - 5 | 514 | 130 | |
GenBank mRNA | AK290765.1 | - | 3 - 4 | 238 | 130 |
AK303565.1 | - | 3 - 4 | 234 | 130 | |
BC002577.2 | - | 3 - 4 | 216 | 130 | |
BC008472.1 | - | 3 - 4 | 197 | 130 | |
BC009576.1 | - | 3 - 4 | 205 | 130 | |
BC015105.1 | - | 3 - 4 | 231 | 130 | |
BC015356.1 | - | 3 - 4 | 200 | 130 | |
BC022372.1 | - | 3 - 4 | 184 | 130 | |
D00759.1 | - | 3 - 4 | 192 | 130 | |
FJ224300.1 | - | 4 - 5 | 496 | 130 | |
M64992.1 | - | 4 - 5 | 496 | 130 | |
X61969.1 | - | 3 - 4 | 256 | 130 |