Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM66
Entrez Gene ID
9866
Gene Name
tripartite motif containing 66
Gene Aliases
C11orf29, TIF1D, TIF1DELTA
Chromosome Location
Chr.11: 8612037 - 8683217 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014818.1 NP_055633.1 9 - 10 1908 139
XM_006718397.2 XP_006718460.1 14 - 15 2753 139
XM_006718398.3 XP_006718461.1 10 - 11 3365 139
XM_011520504.1 XP_011518806.1 15 - 16 2796 139
XM_011520507.2 XP_011518809.1 14 - 15 2717 139
XM_011520508.1 XP_011518810.1 14 - 15 2753 139
XM_011520509.1 XP_011518811.1 14 - 15 2753 139
XM_011520510.1 XP_011518812.1 14 - 15 2753 139
XM_011520511.1 XP_011518813.1 14 - 15 2642 139
XM_011520512.1 XP_011518814.1 10 - 11 2017 139
XM_011520513.1 XP_011518815.1 10 - 11 2015 139
XM_011520514.2 XP_011518816.1 10 - 11 1971 139
XM_011520515.1 XP_011518817.1 11 - 12 2184 139
XM_011520516.1 XP_011518818.1 11 - 12 2184 139
XM_011520517.2 XP_011518819.1 11 - 12 3506 139
XM_011520518.1 XP_011518820.1 11 - 12 2154 139
XM_011520519.1 XP_011518821.1 10 - 11 2042 139
XM_011520522.2 XP_011518824.1 9 - 10 3220 139
XM_011520523.2 XP_011518825.1 10 - 11 4102 139
XM_011520524.1 XP_011518826.1 14 - 15 2753 139
XM_011520525.1 XP_011518827.1 14 - 15 2753 139
XM_011520526.2 XP_011518828.1 2 - 3 1071 139
XM_011520527.1 XP_011518829.1 2 - 3 339 139
XM_017018629.1 XP_016874118.1 14 - 15 3146 139
XM_017018630.1 XP_016874119.1 9 - 10 2108 139
XM_017018631.1 XP_016874120.1 9 - 10 2260 139
GenBank mRNA AB002296.2 - 9 - 10 1908 139

View Full Product Details