The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012335.3 | NP_036467.2 | 17 - 18 | 1941 | 80 |
XM_011528027.1 | XP_011526329.1 | 16 - 17 | 1855 | 80 | |
XM_017026821.1 | XP_016882310.1 | 17 - 18 | 1929 | 80 | |
GenBank mRNA | AB210037.1 | - | 17 - 18 | 1940 | 80 |
AB290178.1 | - | 17 - 18 | 2071 | 80 | |
AK131106.1 | - | 17 - 18 | 1940 | 80 | |
BC028071.1 | - | 17 - 18 | 1917 | 80 |