The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012335.3 | NP_036467.2 | 19 - 20 | 2181 | 85 |
XM_011528024.1 | XP_011526326.1 | 19 - 20 | 2244 | 85 | |
XM_011528025.1 | XP_011526327.1 | 19 - 20 | 2232 | 85 | |
XM_011528026.1 | XP_011526328.1 | 18 - 19 | 2157 | 85 | |
XM_011528027.1 | XP_011526329.1 | 18 - 19 | 2109 | 85 | |
XM_017026821.1 | XP_016882310.1 | 19 - 20 | 2169 | 85 | |
GenBank mRNA | AB210037.1 | - | 19 - 20 | 2180 | 85 |
AB290178.1 | - | 19 - 20 | 2311 | 85 | |
AJ310570.1 | - | 19 - 20 | 2083 | 85 | |
AK131106.1 | - | 19 - 20 | 2180 | 85 | |
BC028071.1 | - | 19 - 20 | 2157 | 85 | |
X98411.1 | - | 10 - 11 | 1092 | 85 |