Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYO1F
Entrez Gene ID
4542
Gene Name
myosin IF
Gene Aliases
-
Chromosome Location
Chr.19: 8520790 - 8577447 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012335.3 NP_036467.2 26 - 27 3192 132
XM_011528024.1 XP_011526326.1 26 - 27 3255 132
XM_011528025.1 XP_011526327.1 26 - 27 3243 132
XM_011528026.1 XP_011526328.1 25 - 26 3168 132
XM_011528027.1 XP_011526329.1 25 - 26 3120 132
XM_017026821.1 XP_016882310.1 26 - 27 3180 132
GenBank mRNA AB210037.1 - 26 - 27 3191 132
AB290178.1 - 26 - 27 3322 132
AJ310570.1 - 26 - 27 3094 132
AK131106.1 - 26 - 27 3193 132
BC028071.1 - 26 - 27 3168 132
X98411.1 - 17 - 18 129

View Full Product Details