The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_174922.4 | NP_777582.4 | 7 - 8 | 874 | 82 |
XM_006716527.2 | XP_006716590.1 | 7 - 8 | 864 | 82 | |
XM_011516907.2 | XP_011515209.1 | 7 - 8 | 1587 | 82 | |
XM_011516909.1 | XP_011515211.1 | 7 - 8 | 874 | 82 | |
XM_011516910.1 | XP_011515212.1 | 8 - 9 | 897 | 82 | |
XM_017013174.1 | XP_016868663.1 | 7 - 8 | 912 | 82 | |
GenBank mRNA | AK092773.1 | - | 7 - 8 | 837 | 82 |
BC101659.1 | - | 7 - 8 | 814 | 82 | |
BC143326.1 | - | 6 - 7 | 641 | 82 |