Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHG6
Entrez Gene ID
55200
Gene Name
pleckstrin homology and RhoGEF domain containing G6
Gene Aliases
MyoGEF
Chromosome Location
Chr.12: 6310436 - 6328506 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144856.1 NP_001138328.1 3 - 4 521 91
NM_001144857.1 NP_001138329.1 2 - 3 689 91
NM_018173.3 NP_060643.2 3 - 4 439 91
XM_005253704.4 XP_005253761.1 3 - 4 536 91
XM_006718985.3 XP_006719048.1 3 - 4 536 91
XM_011520967.2 XP_011519269.1 3 - 4 411 91
XM_017019555.1 XP_016875044.1 3 - 4 536 91
GenBank mRNA AK001527.1 - 3 - 4 440 91
AK023741.1 - 3 - 4 521 91
AK095373.1 - 2 - 3 689 91
BC104751.1 - 3 - 4 375 91
BC105961.1 - 3 - 4 375 91
BC109095.2 - 3 - 4 375 91

View Full Product Details