Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANGPTL7
Entrez Gene ID
10218
Gene Name
angiopoietin like 7
Gene Aliases
AngX, CDT6, dJ647M16.1
Chromosome Location
Chr.1: 11189289 - 11195981 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021146.3 NP_066969.1 2 - 3 769 70
XM_017000004.1 XP_016855493.1 2 - 3 734 70
GenBank mRNA AB593026.1 - 2 - 3 703 70
AB593027.1 - 2 - 3 770 70
AK313716.1 - 2 - 3 735 70
AY358301.1 - 2 - 3 692 70
BC001881.1 - 2 - 3 717 70
BT009802.1 - 2 - 3 478 70
Y16132.1 - 2 - 3 485 70

View Full Product Details