Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TAS1R2
Entrez Gene ID
80834
Gene Name
taste 1 receptor member 2
Gene Aliases
GPR71, T1R2, TR2
Chromosome Location
Chr.1: 18839599 - 18859661 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152232.2 NP_689418.2 5 - 6 1592 56

View Full Product Details