Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITIH2
Entrez Gene ID
3698
Gene Name
inter-alpha-trypsin inhibitor heavy chain 2
Gene Aliases
H2P, SHAP
Chromosome Location
Chr.10: 7703273 - 7749520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002216.2 NP_002207.2 12 - 13 1624 58
GenBank mRNA AK225277.1 - 14 - 15 1487 58
AK225329.1 - 12 - 13 1529 58
AK291379.1 - 12 - 13 1572 58
AK297334.1 - 7 - 8 815 58
BC132685.1 - 12 - 13 1511 58
CB154170.1 - 3 - 4 397 58
M18193.1 - 2 - 3 315 58
M33033.1 - 2 - 3 313 58
X07173.1 - 12 - 13 1525 58

View Full Product Details