Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITIH2
Entrez Gene ID
3698
Gene Name
inter-alpha-trypsin inhibitor heavy chain 2
Gene Aliases
H2P, SHAP
Chromosome Location
Chr.10: 7703273 - 7749520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002216.2 NP_002207.2 17 - 18 2367 80
GenBank mRNA AK225277.1 - 19 - 20 2230 80
AK225329.1 - 17 - 18 2272 80
AK291379.1 - 17 - 18 2315 80
AK297334.1 - 12 - 13 1558 80
BC132685.1 - 17 - 18 2254 80
X07173.1 - 17 - 18 2268 80

View Full Product Details