Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITIH2
Entrez Gene ID
3698
Gene Name
inter-alpha-trypsin inhibitor heavy chain 2
Gene Aliases
H2P, SHAP
Chromosome Location
Chr.10: 7703273 - 7749520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002216.2 NP_002207.2 2 - 3 318 137
GenBank mRNA AK225277.1 - 2 - 3 181 137
AK225329.1 - 2 - 3 223 137
AK291379.1 - 2 - 3 266 137
BC132685.1 - 2 - 3 205 137
X07173.1 - 2 - 3 219 137

View Full Product Details