Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MADD
Entrez Gene ID
8567
Gene Name
MAP kinase activating death domain
Gene Aliases
DENN, IG20, RAB3GEP
Chromosome Location
Chr.11: 47269376 - 47330031 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135943.1 NP_001129415.1 5 - 6 1310 76
NM_001135944.1 NP_001129416.1 5 - 6 1286 76
NM_003682.3 NP_003673.3 5 - 6 1286 76
NM_130470.2 NP_569826.2 5 - 6 1452 76
NM_130471.2 NP_569827.2 5 - 6 1286 76
NM_130472.2 NP_569828.2 5 - 6 1286 76
NM_130473.2 NP_569829.2 5 - 6 1286 76
NM_130474.2 NP_569830.2 5 - 6 1286 76
NM_130475.2 NP_569831.1 5 - 6 1310 76
NM_130476.2 NP_569832.2 5 - 6 1310 76
XM_005253189.2 XP_005253246.1 5 - 6 1310 76
XM_005253196.2 XP_005253253.1 5 - 6 1310 76
XM_005253199.2 XP_005253256.1 5 - 6 1310 76
XM_005253200.2 XP_005253257.1 5 - 6 1310 76
XM_005253201.2 XP_005253258.1 5 - 6 1310 76
XM_005253203.2 XP_005253260.1 5 - 6 1310 76
XM_005253204.2 XP_005253261.1 5 - 6 1310 76
XM_005253205.1 XP_005253262.1 5 - 6 1310 76
XM_017018478.1 XP_016873967.1 5 - 6 1310 76
XM_017018479.1 XP_016873968.1 5 - 6 1310 76
XM_017018480.1 XP_016873969.1 5 - 6 1310 76
XM_017018481.1 XP_016873970.1 5 - 6 1310 76
XM_017018482.1 XP_016873971.1 5 - 6 1310 76
XM_017018483.1 XP_016873972.1 5 - 6 1310 76
XM_017018484.1 XP_016873973.1 5 - 6 1310 76
XM_017018485.1 XP_016873974.1 5 - 6 1271 76
XM_017018486.1 XP_016873975.1 5 - 6 1310 76
XM_017018487.1 XP_016873976.1 5 - 6 1310 76
XM_017018488.1 XP_016873977.1 5 - 6 1310 76
XM_017018489.1 XP_016873978.1 5 - 6 1271 76
XM_017018490.1 XP_016873979.1 5 - 6 1297 76
XM_017018491.1 XP_016873980.1 5 - 6 1266 76
XM_017018492.1 XP_016873981.1 5 - 6 1310 76
XM_017018493.1 XP_016873982.1 5 - 6 1297 76
XM_017018494.1 XP_016873983.1 5 - 6 1310 76
XM_017018495.1 XP_016873984.1 5 - 6 1310 76
XM_017018496.1 XP_016873985.1 5 - 6 1266 76
XM_017018497.1 XP_016873986.1 5 - 6 1271 76
XM_017018498.1 XP_016873987.1 5 - 6 1310 76
XM_017018499.1 XP_016873988.1 5 - 6 1279 76
XM_017018500.1 XP_016873989.1 5 - 6 1310 76
XM_017018501.1 XP_016873990.1 5 - 6 1266 76
XM_017018502.1 XP_016873991.1 5 - 6 1279 76
XM_017018503.1 XP_016873992.1 5 - 6 1266 76
XM_017018504.1 XP_016873993.1 5 - 6 1279 76
XM_017018505.1 XP_016873994.1 5 - 6 1266 76
XM_017018506.1 XP_016873995.1 5 - 6 1271 76
XM_017018507.1 XP_016873996.1 5 - 6 1297 76
XM_017018508.1 XP_016873997.1 5 - 6 1310 76
XM_017018509.1 XP_016873998.1 5 - 6 1266 76
XM_017018510.1 XP_016873999.1 5 - 6 1310 76
GenBank mRNA AB002356.1 - 5 - 6 1282 76
AF440100.1 - 5 - 6 1260 76
AF440101.1 - 5 - 6 1260 76
AF440102.1 - 5 - 6 1260 76
AF440103.1 - 5 - 6 1260 76
AF440434.1 - 5 - 6 1260 76
AK292442.1 - 5 - 6 1250 76
BC040484.1 - 5 - 6 1294 76
U44953.1 - 5 - 6 1274 76
U77352.1 - 5 - 6 1420 76

View Full Product Details