Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C11orf49
Entrez Gene ID
79096
Gene Name
chromosome 11 open reading frame 49
Gene Aliases
-
Chromosome Location
Chr.11: 46936524 - 47164381 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003676.2 NP_001003676.1 2 - 3 258 93
NM_001003677.2 NP_001003677.1 2 - 3 258 93
NM_001003678.2 NP_001003678.1 2 - 3 258 93
NM_024113.4 NP_077018.1 2 - 3 258 93
NR_103471.1 2 - 3 258 93
XM_006718315.1 XP_006718378.1 2 - 3 249 93
XM_011520365.2 XP_011518667.1 2 - 3 249 93
XM_017018276.1 XP_016873765.1 2 - 3 249 93
XM_017018278.1 XP_016873767.1 2 - 3 250 93
XM_017018279.1 XP_016873768.1 2 - 3 250 93
XM_017018280.1 XP_016873769.1 2 - 3 186 93
XM_017018282.1 XP_016873771.1 2 - 3 250 93
GenBank mRNA AK025863.1 - 2 - 3 167 93
AK315639.1 - 2 - 3 206 93
AK316318.1 - 2 - 3 250 93
AL136575.1 - 2 - 3 177 93
BC001860.2 - 2 - 3 204 93
BC009455.1 - 2 - 3 179 93
BC013988.2 - 2 - 3 169 93
BC052291.1 - 2 - 3 171 93
CR457351.1 - 2 - 3 141 93
DB027437.1 - 2 - 3 249 93

View Full Product Details