Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C11orf49
Entrez Gene ID
79096
Gene Name
chromosome 11 open reading frame 49
Gene Aliases
-
Chromosome Location
Chr.11: 46936524 - 47164381 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003676.2 NP_001003676.1 3 - 4 395 83
NM_001003677.2 NP_001003677.1 3 - 4 395 83
NM_001003678.2 NP_001003678.1 3 - 4 395 83
NM_001278222.1 NP_001265151.1 2 - 3 292 83
NM_024113.4 NP_077018.1 3 - 4 395 83
XM_006718315.1 XP_006718378.1 3 - 4 386 83
XM_011520364.2 XP_011518666.1 2 - 3 457 83
XM_011520365.2 XP_011518667.1 3 - 4 386 83
XM_017018276.1 XP_016873765.1 3 - 4 386 83
GenBank mRNA AK300813.1 - 2 - 3 202 83
AK315639.1 - 3 - 4 343 83
AL136575.1 - 3 - 4 314 83
BC001860.2 - 3 - 4 341 83
BC009455.1 - 3 - 4 316 83
BC013988.2 - 3 - 4 306 83
BC052291.1 - 3 - 4 308 83
CR457351.1 - 3 - 4 278 83
DB027437.1 - 3 - 4 386 83

View Full Product Details