Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C11orf49
Entrez Gene ID
79096
Gene Name
chromosome 11 open reading frame 49
Gene Aliases
-
Chromosome Location
Chr.11: 46936524 - 47164381 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001003676.2 NP_001003676.1 6 - 7 72
NM_001003677.2 NP_001003677.1 6 - 7 72
NM_001003678.2 NP_001003678.1 6 - 7 72
NM_001278222.1 NP_001265151.1 5 - 6 72
NM_024113.4 NP_077018.1 6 - 7 72
NR_103471.1 7 - 8 72
NR_103472.1 5 - 6 72
XM_006718315.1 XP_006718378.1 6 - 7 72
XM_011520364.2 XP_011518666.1 5 - 6 72
XM_011520365.2 XP_011518667.1 6 - 7 72
XM_017018276.1 XP_016873765.1 6 - 7 72
XM_017018277.1 XP_016873766.1 5 - 6 72
XM_017018278.1 XP_016873767.1 7 - 8 72
XM_017018279.1 XP_016873768.1 7 - 8 72
XM_017018280.1 XP_016873769.1 8 - 9 72
XM_017018281.1 XP_016873770.1 7 - 8 72
XM_017018282.1 XP_016873771.1 7 - 8 72
GenBank mRNA AK025863.1 - 6 - 7 72
AK293609.1 - 5 - 6 72
AK300813.1 - 5 - 6 72
AK315639.1 - 6 - 7 72
AK316318.1 - 7 - 8 72
AL136575.1 - 6 - 7 72
BC001860.2 - 6 - 7 72
BC009455.1 - 6 - 7 72
BC013988.2 - 6 - 7 72
BC052291.1 - 6 - 7 72
CK724853.1 - 3 - 4 480 72
CR457351.1 - 6 - 7 72

View Full Product Details