The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175732.2 | NP_783859.1 | 1 - 2 | 365 | 53 |
GenBank mRNA | AK294640.1 | - | 1 - 2 | 196 | 53 |
AY333987.1 | - | 1 - 2 | 172 | 53 | |
BC020242.1 | - | 1 - 2 | 194 | 53 | |
DA476984.1 | - | 1 - 2 | 221 | 53 | |
DR004030.1 | - | 1 - 2 | 223 | 53 |