The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006345.3 | NP_006336.3 | 13 - 14 | 1293 | 124 |
XM_017007654.1 | XP_016863143.1 | 13 - 14 | 1327 | 124 | |
GenBank mRNA | AF006621.2 | - | 13 - 14 | 1267 | 124 |
AK225470.1 | - | 13 - 14 | 1230 | 124 | |
AK299917.1 | - | 14 - 15 | 1387 | 124 | |
AK312838.1 | - | 13 - 14 | 1293 | 124 | |
AY319413.1 | - | 14 - 15 | 1172 | 124 | |
AY594282.1 | - | 13 - 14 | 1147 | 124 | |
BC000240.1 | - | 13 - 14 | 1255 | 124 | |
BC007732.2 | - | 13 - 14 | 1254 | 124 | |
BC016949.1 | - | 13 - 14 | 1272 | 124 | |
BC022981.1 | - | 9 - 10 | 684 | 124 | |
KU178526.1 | - | 13 - 14 | 1147 | 124 | |
KU178527.1 | - | 13 - 14 | 1212 | 124 |