The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004535.2 | NP_004526.1 | 18 - 19 | 3037 | 62 |
GenBank mRNA | AB020642.1 | - | 18 - 19 | 3037 | 62 |
BC053638.1 | - | 12 - 13 | 1849 | 62 | |
M96980.1 | - | 12 - 13 | 1485 | 62 |