The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004535.2 | NP_004526.1 | 20 - 21 | 3354 | 68 |
GenBank mRNA | AB020642.1 | - | 20 - 21 | 3354 | 68 |
BC053638.1 | - | 14 - 15 | 2166 | 68 | |
M96980.1 | - | 14 - 15 | 1802 | 68 |