The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001118.4 | NP_001109.2 | 8 - 9 | 825 | 86 |
NM_001199635.1 | NP_001186564.1 | 8 - 9 | 825 | 86 | |
NM_001199636.1 | NP_001186565.1 | 8 - 9 | 825 | 86 | |
NM_001199637.1 | NP_001186566.1 | 6 - 7 | 762 | 86 | |
XM_005249618.4 | XP_005249675.1 | 5 - 6 | 771 | 86 | |
XM_006715645.2 | XP_006715708.1 | 8 - 9 | 825 | 86 | |
XM_017011736.1 | XP_016867225.1 | 7 - 8 | 804 | 86 | |
XM_017011737.1 | XP_016867226.1 | 6 - 7 | 762 | 86 | |
XM_017011738.1 | XP_016867227.1 | 5 - 6 | 774 | 86 | |
GenBank mRNA | AK226138.1 | - | 6 - 7 | 696 | 86 |
AK290046.1 | - | 8 - 9 | 776 | 86 | |
AY366498.1 | - | 7 - 8 | 536 | 86 | |
BC117116.1 | - | 7 - 8 | 610 | 86 | |
BC136267.1 | - | 7 - 8 | 610 | 86 | |
BC143679.1 | - | 7 - 8 | 610 | 86 |