The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243835.1 | NP_001230764.1 | 23 - 24 | 2488 | 87 |
NM_003151.3 | NP_003142.1 | 23 - 24 | 2539 | 87 | |
XM_006712719.3 | XP_006712782.1 | 23 - 24 | 2534 | 87 | |
XM_011511705.2 | XP_011510007.1 | 26 - 27 | 2685 | 87 | |
GenBank mRNA | AK299976.1 | - | 20 - 21 | 2020 | 87 |
BC031212.1 | - | 23 - 24 | 2460 | 87 | |
L78440.1 | - | 23 - 24 | 2305 | 87 |