The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005323.3 | NP_005314.2 | 1 - 1 | 443 | 151 |
GenBank mRNA | BC069517.1 | - | 1 - 1 | 443 | 151 |
BC130350.1 | - | 1 - 1 | 436 | 151 |