Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TTC24
Entrez Gene ID
164118
Gene Name
tetratricopeptide repeat domain 24
Gene Aliases
-
Chromosome Location
Chr.1: 156579727 - 156587717 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105669.2 NP_001099139.2 9 - 10 1608 64
XM_011509253.2 XP_011507555.1 8 - 9 990 64
XM_011509254.2 XP_011507556.1 8 - 9 987 64
XM_017000497.1 XP_016855986.1 6 - 7 612 64
GenBank mRNA AW628680.1 - 2 - 3 64

View Full Product Details