The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181807.3 | NP_861523.2 | 3 - 4 | 393 | 90 |
GenBank mRNA | AY247970.1 | - | 3 - 4 | 357 | 90 |
DB090201.1 | - | 3 - 4 | 393 | 90 |