Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ITPKB
Entrez Gene ID
3707
Gene Name
inositol-trisphosphate 3-kinase B
Gene Aliases
IP3-3KB, IP3K, IP3K-B, IP3KB, PIG37
Chromosome Location
Chr.1: 226631690 - 226739327 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002221.3 NP_002212.3 5 - 6 2788 106
GenBank mRNA AJ242780.1 - 4 - 5 2505 106
AK289700.1 - 4 - 5 1234 106
AK313801.1 - 4 - 5 1253 106
AY513281.1 - 5 - 6 1029 106
X57206.1 - 4 - 5 1127 106

View Full Product Details