Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIH1D2
Entrez Gene ID
120379
Gene Name
PIH1 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.11: 112052161 - 112074288 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138789.3 NP_620144.1 5 - 6 1033 129
GenBank mRNA BC019238.2 - 4 - 5 842 129

View Full Product Details