The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243963.1 | NP_001230892.1 | 17 - 18 | 1940 | 79 |
NM_020871.3 | NP_065922.3 | 18 - 19 | 1991 | 79 | |
XM_006724724.3 | XP_006724787.2 | 18 - 19 | 1973 | 79 | |
GenBank mRNA | AB040928.1 | - | 14 - 15 | 1172 | 79 |
AK022128.1 | - | 10 - 11 | 747 | 79 | |
BC125223.1 | - | 6 - 7 | 398 | 79 | |
BC125224.1 | - | 17 - 18 | 1933 | 79 | |
BX647193.1 | - | 18 - 19 | 1991 | 79 |