The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001243963.1 | NP_001230892.1 | 18 - 19 | 2079 | 75 |
NM_020871.3 | NP_065922.3 | 19 - 20 | 2130 | 75 | |
XM_006724724.3 | XP_006724787.2 | 19 - 20 | 2112 | 75 | |
GenBank mRNA | AB040928.1 | - | 15 - 16 | 1311 | 75 |
AK022128.1 | - | 11 - 12 | 886 | 75 | |
BC125223.1 | - | 7 - 8 | 537 | 75 | |
BX647193.1 | - | 19 - 20 | 2130 | 75 |