The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130107.1 | NP_001123579.1 | 10 - 11 | 1618 | 78 |
NM_005552.4 | NP_005543.2 | 10 - 11 | 1618 | 78 | |
NM_182923.3 | NP_891553.2 | 10 - 11 | 1618 | 78 | |
GenBank mRNA | AB451299.1 | - | 9 - 10 | 1298 | 78 |
AB451431.1 | - | 9 - 10 | 1298 | 78 | |
AK294508.1 | - | 10 - 11 | 1603 | 78 | |
AY180164.2 | - | 10 - 11 | 1565 | 78 | |
BC008881.2 | - | 10 - 11 | 1579 | 78 | |
L04733.1 | - | 10 - 11 | 1574 | 78 |