The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130107.1 | NP_001123579.1 | 6 - 7 | 1194 | 110 |
NM_005552.4 | NP_005543.2 | 6 - 7 | 1194 | 110 | |
NM_182923.3 | NP_891553.2 | 6 - 7 | 1194 | 110 | |
GenBank mRNA | AB451299.1 | - | 5 - 6 | 874 | 110 |
AB451431.1 | - | 5 - 6 | 874 | 110 | |
AF527736.1 | - | 6 - 7 | 1144 | 110 | |
AK294508.1 | - | 6 - 7 | 1179 | 110 | |
AY180164.2 | - | 6 - 7 | 1141 | 110 | |
BC008881.2 | - | 6 - 7 | 1155 | 110 | |
L04733.1 | - | 6 - 7 | 1150 | 110 |