The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270965.1 | NP_001257894.1 | 13 - 14 | 2533 | 68 |
NM_001270966.1 | NP_001257895.1 | 13 - 14 | 2054 | 68 | |
NM_002779.4 | NP_002770.3 | 12 - 13 | 2857 | 68 | |
NR_073110.1 | 3 - 4 | 629 | 68 | ||
XM_011539968.1 | XP_011538270.1 | 13 - 14 | 2539 | 68 | |
XM_011539969.2 | XP_011538271.1 | 13 - 14 | 2556 | 68 | |
XM_017016433.1 | XP_016871922.1 | 13 - 14 | 2550 | 68 | |
XM_017016434.1 | XP_016871923.1 | 12 - 13 | 2477 | 68 | |
GenBank mRNA | AB095931.1 | - | 12 - 13 | 2857 | 68 |
BC039259.1 | - | 13 - 14 | 2041 | 68 | |
BC142643.1 | - | 13 - 14 | 2533 | 68 | |
BC142689.1 | - | 13 - 14 | 2533 | 68 | |
BM920983.1 | - | - | 360 | 68 |