The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077494.3 | NP_001070962.1 | 16 - 17 | 2068 | 91 |
NM_001261403.2 | NP_001248332.1 | 15 - 16 | 2163 | 91 | |
NM_001288724.1 | NP_001275653.1 | 16 - 17 | 2362 | 91 | |
NM_001322934.1 | NP_001309863.1 | 16 - 17 | 2072 | 91 | |
NM_001322935.1 | NP_001309864.1 | 15 - 16 | 2236 | 91 | |
NM_002502.5 | NP_002493.3 | 16 - 17 | 2072 | 91 | |
XM_011539830.2 | XP_011538132.1 | 11 - 12 | 1426 | 91 | |
XM_011539831.2 | XP_011538133.1 | 11 - 12 | 1423 | 91 | |
XM_017016278.1 | XP_016871767.1 | 16 - 17 | 2357 | 91 | |
GenBank mRNA | AK098415.1 | - | 6 - 7 | 946 | 91 |
AK292654.1 | - | 15 - 16 | 2016 | 91 | |
BC002844.2 | - | 16 - 17 | 1899 | 91 | |
BT009769.1 | - | 15 - 16 | 1799 | 91 | |
S76638.1 | - | 16 - 17 | 2049 | 91 | |
U09609.2 | - | 15 - 16 | 1799 | 91 | |
X61498.1 | - | 16 - 17 | 1959 | 91 |