The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077494.3 | NP_001070962.1 | 18 - 19 | 2343 | 103 |
NM_001261403.2 | NP_001248332.1 | 17 - 18 | 2438 | 103 | |
NM_001288724.1 | NP_001275653.1 | 18 - 19 | 2637 | 103 | |
NM_001322934.1 | NP_001309863.1 | 18 - 19 | 2347 | 103 | |
NM_001322935.1 | NP_001309864.1 | 17 - 18 | 2511 | 103 | |
NM_002502.5 | NP_002493.3 | 18 - 19 | 2347 | 103 | |
XM_011539830.2 | XP_011538132.1 | 13 - 14 | 1701 | 103 | |
XM_011539831.2 | XP_011538133.1 | 13 - 14 | 1698 | 103 | |
XM_017016278.1 | XP_016871767.1 | 18 - 19 | 2632 | 103 | |
GenBank mRNA | AK098415.1 | - | 8 - 9 | 1220 | 103 |
AK292654.1 | - | 17 - 18 | 2291 | 103 | |
BC002844.2 | - | 18 - 19 | 2174 | 103 | |
BT009769.1 | - | 17 - 18 | 2074 | 103 | |
S76638.1 | - | 18 - 19 | 2324 | 103 | |
X61498.1 | - | 18 - 19 | 2234 | 103 |