Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NFKB2
Entrez Gene ID
4791
Gene Name
nuclear factor kappa B subunit 2
Gene Aliases
CVID10, H2TF1, LYT-10, LYT10, NF-kB2, p100, p49/p100, p52
Chromosome Location
Chr.10: 102394110 - 102402529 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001288724.1 NP_001275653.1 1 - 2 495 70
NM_001322935.1 NP_001309864.1 1 - 2 495 70
GenBank mRNA BY799958.2 - 1 - 2 133 70

View Full Product Details