The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077494.3 | NP_001070962.1 | 4 - 5 | 417 | 63 |
NM_001261403.2 | NP_001248332.1 | 3 - 4 | 512 | 63 | |
NM_001288724.1 | NP_001275653.1 | 4 - 5 | 711 | 63 | |
NM_001322934.1 | NP_001309863.1 | 4 - 5 | 421 | 63 | |
NM_001322935.1 | NP_001309864.1 | 4 - 5 | 711 | 63 | |
NM_002502.5 | NP_002493.3 | 4 - 5 | 421 | 63 | |
XM_017016278.1 | XP_016871767.1 | 4 - 5 | 706 | 63 | |
GenBank mRNA | AK292654.1 | - | 3 - 4 | 365 | 63 |
BC002844.2 | - | 4 - 5 | 248 | 63 | |
BT009769.1 | - | 3 - 4 | 148 | 63 | |
BY799958.2 | - | 4 - 5 | 349 | 63 | |
DA349980.1 | - | 3 - 4 | 433 | 63 | |
DA408461.1 | - | 4 - 5 | 348 | 63 | |
DA711063.1 | - | 4 - 5 | 415 | 63 | |
S76638.1 | - | 4 - 5 | 398 | 63 | |
U09609.2 | - | 3 - 4 | 148 | 63 | |
X61498.1 | - | 4 - 5 | 311 | 63 | |
X61499.1 | - | 4 - 5 | 311 | 63 |