The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000089.3 | NP_000080.2 | 48 - 49 | 3743 | 85 |
GenBank mRNA | AK226074.1 | - | 48 - 49 | 3394 | 85 |
BC042586.1 | - | 48 - 49 | 3390 | 85 | |
BC054498.1 | - | 48 - 49 | 3372 | 85 | |
J03464.1 | - | 48 - 49 | 3741 | 85 | |
KU177988.1 | - | 48 - 49 | 3272 | 85 | |
V00503.1 | - | 17 - 18 | 1407 | 85 | |
X55525.1 | - | 5 - 6 | 397 | 85 | |
Z74616.1 | - | 48 - 49 | 3411 | 85 |