Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PGA5
Entrez Gene ID
5222
Gene Name
pepsinogen 5, group I (pepsinogen A)
Gene Aliases
Pg5
Chromosome Location
Chr.11: 61241176 - 61251444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014224.4 NP_055039.1 6 - 7 820 97
GenBank mRNA BC029055.1 - 6 - 7 799 97
BC146999.1 - 6 - 7 820 97
BC171889.1 - 6 - 7 820 97
BC171897.1 - 6 - 7 820 97

Gene Symbol
PGA3
Entrez Gene ID
643834
Gene Name
pepsinogen 3, group I (pepsinogen A)
Gene Aliases
-
Chromosome Location
Chr.11: 61203513 - 61212869 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079807.3 NP_001073275.1 6 - 7 821 97
GenBank mRNA AK125628.1 - 6 - 7 835 97
AK225669.1 - 6 - 7 819 97
AK225674.1 - 6 - 7 847 97
AK225679.1 - 6 - 7 819 97
AK301286.1 - 7 - 8 692 97
AK312941.1 - 6 - 7 821 97
BC171815.1 - 6 - 7 815 97

Gene Symbol
PGA4
Entrez Gene ID
643847
Gene Name
pepsinogen 4, group I (pepsinogen A)
Gene Aliases
-
Chromosome Location
Chr.11: 61222349 - 61231695 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079808.3 NP_001073276.1 6 - 7 821 97
GenBank mRNA AK291864.1 - 6 - 7 819 97
BC150659.1 - 6 - 7 815 97
BC171808.1 - 6 - 7 815 97
BC171814.1 - 6 - 7 815 97
BC171910.1 - 6 - 7 815 97
BC171920.1 - 6 - 7 815 97

View Full Product Details