Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACTR3
Entrez Gene ID
10096
Gene Name
ARP3 actin related protein 3 homolog
Gene Aliases
ARP3
Chromosome Location
Chr.2: 113889934 - 113961554 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005721.4 NP_005712.1 1 - 2 384 102
NR_102318.1 1 - 2 384 102
GenBank mRNA AF006083.1 - 1 - 2 210 102
AF127773.1 - 1 - 2 253 102
AK300030.1 - 1 - 2 208 102
AK303988.1 - 1 - 2 221 102
AK312659.1 - 1 - 2 184 102
BC044590.1 - 1 - 2 358 102
DB453995.1 - 1 - 2 384 102

View Full Product Details