The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001415.3 | NP_001406.1 | 1 - 2 | 87 | 85 |
GenBank mRNA | AB451230.1 | - | 1 - 2 | 66 | 85 |
AB451353.1 | - | 1 - 2 | 66 | 85 | |
AK222577.1 | - | 1 - 2 | 79 | 85 | |
AK290397.1 | - | 1 - 2 | 79 | 85 | |
AK312247.1 | - | 1 - 2 | 79 | 85 | |
BC019906.1 | - | 1 - 2 | 87 | 85 | |
L19161.1 | - | 1 - 2 | 66 | 85 |