The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007269.2 | NP_009200.2 | 16 - 17 | 1521 | 89 |
GenBank mRNA | AF032922.1 | - | 16 - 17 | 1503 | 89 |
AK223170.1 | - | 16 - 17 | 1500 | 89 | |
AK290134.1 | - | 16 - 17 | 1505 | 89 | |
AK291322.1 | - | 16 - 17 | 1501 | 89 | |
AK312910.1 | - | 16 - 17 | 1498 | 89 | |
BC038099.2 | - | 16 - 17 | 1460 | 89 | |
BC047764.1 | - | 16 - 17 | 1496 | 89 | |
D63506.1 | - | 16 - 17 | 1497 | 89 |